EMAIL THIS PAGE TO A FRIEND
Toxicology letters
Cell uptake and oral absorption of titanium dioxide nanoparticles.
PMID 24793716
Abstract
Large efforts are invested on the development of in vitro tests to evaluate nanomaterial (NM) toxicity. In order to assess the relevance of the adverse effects identified in in vitro toxicity tests a thorough understanding of the biokinetics of NMs is critical. We used different in vitro and in vivo test methods to evaluate cell uptake and oral absorption of titanium dioxide nanoparticles (TiO2 NPs). These NPs were readily uptaken by A549 cells (carcinomic human alveolar basal epithelial cells) in vitro. Such rapid uptake contrasted with a very low oral absorption in a differentiated Caco-2 monolayer system (human epithelial colorectal adenocarcinoma cells) and after oral gavage administration to rats. In this oral study, no significant increase in the levels of titanium was recorded by ICP-MS in any of the tissues evaluated (including among other: small intestine, Peyer's patches, mesenteric lymph nodes, liver, and spleen). No NPs were observed by TEM in sections of the small intestine, except for several particles in the cytoplasm of a cell from a Peyer's Patch area. The observation of NPs in Peyer's Patch suggests that the Caco-2 monolayer system is likely to underestimate the potential for oral absorption of NPs and that the model could be improved by including M-cells in co-culture.