EMAIL THIS PAGE TO A FRIEND
Brain, behavior, and immunity
The SHP-1 expression is associated with cytokines and psychopathological status in unmedicated first episode schizophrenia patients.
PMID 24793756
Abstract
Recent lines of research have boosted awareness of the immunological facets of schizophrenia. However, associations with protein tyrosine phosphatase regulators have never been reported. The aim of our study was to investigate the expression and promoter status methylation of phosphatase SHP-1, a key negative regulator of the inflammatory process, in Peripheral blood mononuclear cells (PBMCs) of Schizophrenic patients. We enrolled fifty-four (28 men and 26 women) unmedicated first episode subjects (SC) who met DSM-IV and thirty-eight (22 men and 16 women) healthy controls (HC). The SC psychopathological status was assessed using the Positive and Negative Syndrome Scale. We evaluated SHP-1 expression by Quantitative Real-time PCR (qPCR) and Western blotting (WB) methods and promoter status methylation through PCR bisulfate. IKK/NFkB signaling was detected by WB, and medium and plasma levels of pro-inflammatory cytokines (IL-1β, IL-2, and TNF-α) by the ELISA method. SHP-1 was silenced by treating cells with specific siRNA. We found a significantly lower level of SHP-1 gene expression in PBMCs from SC vs. HC, consistently with which the promoter region analyzed presented significant hypermethylation. Silencing of SHP-1 expression induced higher activation of IKK/NF-kB signaling and pro-inflammatory cytokine production in ex vivo PBMCs from both SC and HC. Linear regression among patients generated a model in which SHP-1 expression explained 30% of the clinical negative symptom variance (adjusted R(2)=0.30, ANOVA p<0.001). Our findings are the first to suggest that impairment of SHP-1 expression is involved in the physiopathology of schizophrenia, opening fruitful new avenues for ameliorating treatment at least of negative symptoms.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB2101628
Anti-NR0B2 (ab1) antibody produced in rabbit, affinity isolated antibody
SAB2101629
Anti-NR0B2 (ab2) antibody produced in rabbit, affinity isolated antibody
SAB2101630
Anti-NR0B2 (ab3) antibody produced in rabbit, affinity isolated antibody
SAB2101631
Anti-NR0B2 (ab4) antibody produced in rabbit, affinity isolated antibody
SAB1406664
Anti-NR0B2 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB4300558
Anti-PTPN6 (Ab-536) antibody produced in rabbit, affinity isolated antibody
SAB1406344
Anti-PTPN6 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA001466
Anti-PTPN6 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
GW21487 Anti-PTPNS1 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB1408349
Anti-SIRPA antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2102154
Anti-SIRPA antibody produced in rabbit, affinity isolated antibody
WH0008431M1
Monoclonal Anti-NR0B2 antibody produced in mouse, clone 1A11, purified immunoglobulin, buffered aqueous solution
WH0140885M1
Monoclonal Anti-PTPNS1 antibody produced in mouse, clone 1D10, purified immunoglobulin, buffered aqueous solution
RAB0477
Mouse Tumor Necrosis Factor α ELISA Kit, for serum, plasma and cell culture supernatant
RAB0479
Rat Tumor Necrosis Factor α ELISA Kit, for serum, plasma and cell culture supernatant