EMAIL THIS PAGE TO A FRIEND
JAMA
Idiopathic short stature: a clinical review.
PMID 24794372
Abstract
Approximately 2% of children are defined as having short stature. Deciding when to pursue recombinant human growth hormone therapy to increase adult height is controversial. To review the management of children with idiopathic short stature, including diagnostic evaluation and therapeutic options. Systematic literature search of PubMed, Embase, and the Cochrane Library databases. For height outcome, articles were limited to studies reporting adult height and to systematic reviews. Recombinant human growth hormone therapy of children with idiopathic short stature increases height in some children. The estimated mean gain in adult height is 5.2 cm (2 in). The cost-benefit ratio is controversial. Treatment with growth hormone appears safe in the short term, while data on long-term effects are limited because studies of long-term efficacy were not powered to determine safety. Growth hormone treatment may be considered in some children with idiopathic short stature.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S8648 Somatotropin (STH) from porcine pituitary