EMAIL THIS PAGE TO A FRIEND
The Medical journal of Australia
Rapid eye movement behaviour disorder.
PMID 24794613
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C2385000
Clonazepam, European Pharmacopoeia (EP) Reference Standard
C15H10ClN3O3