EMAIL THIS PAGE TO A FRIEND
Journal of Chromatography A
Identification of effective combinatorial markers for quality standardization of herbal medicines.
PMID 24794938
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D218200
5,5′-Dithiobis(2-nitrobenzoic acid), ReagentPlus®, 99%
C14H8N2O8S2
A5751
Acetylthiocholine iodide, ≥98% (TLC), powder or crystals
C7H16INOS
01480
Acetylthiocholine iodide, ≥99.0% (AT)
C7H16INOS
242381
Benzoic acid, ACS reagent, ≥99.5%
C7H6O2
33045
Benzoic acid, for calorimetrical determination (approx. 26460 J/g)
C7H6O2
06185
Benzoic acid, Standard for quantitative NMR, TraceCERT®
C7H6O2
1055002
Benzoic acid, United States Pharmacopeia (USP) Reference Standard
C7H6O2
33047
Benzoic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9% (alkalimetric)
C7H6O2
18102
Benzoic acid, meets analytical specification of Ph. Eur., BP, USP, FCC, E210, 99.5-100.5% (alkalimetric)
C7H6O2
12356
Benzoic acid, tested according to Ph.Eur.
C7H6O2
PHR1050
Benzoic acid, Pharmaceutical Secondary Standard; Certified Reference Material
C7H6O2
12353
Benzoic acid, certified reference material for titrimetry, certified by BAM according to ISO 17025, ≥99.5%
C7H6O2
47849
Benzoic acid, analytical standard
C7H6O2
427608
Benzoic acid, purified by sublimation, ≥99%
C7H6O2
109479
Benzoic acid, ReagentPlus®, 99%
C7H6O2
W213101
Benzoic acid, ≥99.5%, FCC, FG
C7H6O2
W213128
Benzoic acid, natural, ≥99.5%, FCC, FG
C7H6O2
491861
Benzoic acid-12C7, 99.9 atom % 12C
12C7H6O2
76170
Melting point standard 121-123°C, analytical standard
C7H6O2
73983
Mettler-Toledo Calibration substance ME 18555, Benzoic acid, analytical standard, for the calibration of the thermosystem 900, traceable to primary standards (LGC)
C7H6O2
8S61336
SS Benzoic Acid, 2000 μg/mL in dichloromethane, analytical standard
C7H6O2