EMAIL THIS PAGE TO A FRIEND
The Journal of infectious diseases
Soluble markers of inflammation and coagulation but not T-cell activation predict non-AIDS-defining morbid events during suppressive antiretroviral treatment.
PMID 24795473
Abstract
Defining the association of non-AIDS-defining events with inflammation and immune activation among human immunodeficiency virus (HIV)-infected persons with antiretroviral therapy (ART)-associated virological suppression is critical to identifying interventions to decrease the occurrence of these events. We conducted a case-control study of HIV-infected subjects who had achieved virological suppression within 1 year after ART initiation. Cases were patients who experienced non-AIDS-defining events, defined as myocardial infarction, stroke, non-AIDS-defining cancer, non-AIDS-defining serious bacterial infection, or death. Controls were matched to cases on the basis of age, sex, pre-ART CD4(+) T-cell count, and ART regimen. Peripheral blood mononuclear cells and plasma specimens obtained at the visit before ART initiation (hereafter, baseline), the visit approximately 1 year after ART initiation (hereafter, year 1), and the visit immediately preceding the non-AIDS-defining event (hereafter, pre-event) were analyzed for activated CD4(+) and CD8(+) T cells, plasma interleukin 6 (IL-6) level, soluble tumor necrosis factor receptor I (sTNFR-I) level, sTNFR-II level, soluble CD14 level, kynurenine-to-tryptophan (KT) ratio, and D-dimer level. Conditional logistic regression analysis was used to study the association between biomarkers and outcomes, with adjustment for potential confounders. Higher IL-6 level, sTNFR-I level, sTNFR-II level, KT ratio, and D-dimer level at year 1 were associated with the occurrence of a non-AIDS-defining event. Significant associations were also seen between non-AIDS-defining events and values of these biomarkers in specimens obtained at baseline and the pre-event time points. Effects remained significant after control for confounders. T-cell activation was not significantly related to outcomes. Interventional trials to decrease the incidence of non-AIDS-defining events among HIV-infected persons with virological suppression should consider targeting the pathways represented by these soluble markers. Clinical Trials Registration. NCT00001137.