EMAIL THIS PAGE TO A FRIEND
Interactive Cardiovascular and Thoracic Surgery
Impact of closed minimal extracorporeal circulation on microvascular tissue perfusion during surgical aortic valve replacement: intravital imaging in a prospective randomized study.
PMID 24796334