EMAIL THIS PAGE TO A FRIEND
Nature communications
IL-27 and IL-12 oppose pro-inflammatory IL-23 in CD4+ T cells by inducing Blimp1.
PMID 24796719
Abstract
Central nervous system (CNS) autoimmunity is regulated by the balance of pro-inflammatory cytokines and IL-10. Here we identify the transcriptional regulator Blimp1 as crucial to induce IL-10 in inflammatory T helper cells. Pre-committed Th17 cells respond to IL-27 and IL-12 by upregulating Blimp1 and adopt a Tr-1-like phenotype characterized by IL-10 and IFN-γ production. Accordingly, Blimp1-deficient effector T cells fail to produce IL-10, and deficiency in Tr-1 cell function leads to uncontrolled Th17 cell-driven CNS pathology without the need to stabilize the Th17 phenotype with IL-23. IL-23 counteracts IL-27 and IL-12-mediated effects on Tr-1-development reinforcing the pro-inflammatory phenotype of Th17 cells. Thus, the balance of IL-23 vs IL-12/IL-27 signals into CD4(+) effector T cells determines whether tissue inflammation is perpetuated or resolves.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
50553
α-tri-Calcium phosphate, Reagent for transient & stable DNA transfections
Ca3O8P2
53737
α-tri-Calcium phosphate, puriss. p.a., ≥75% α-phase basis (sinterted powder)
Ca3O8P2
PRS3989
Anti-Blimp-1 (ab1) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
PRS3991
Anti-Blimp-1 (ab2) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
AV54329
Anti-IL4 antibody produced in rabbit, affinity isolated antibody
SAB2103750
Anti-IL4 antibody produced in rabbit, affinity isolated antibody
I7776 Anti-Interleukin-4 antibody produced in goat, affinity isolated antibody, lyophilized powder
I7526 Anti-Interleukin-4 antibody produced in goat, affinity isolated antibody, lyophilized powder
I8784 Anti-Interleukin-4 antibody produced in goat, IgG fraction of antiserum, lyophilized powder
SAB2501379
Anti-PRDM1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1412469
ANTI-PRDM1 antibody produced in mouse, clone 1G6, purified immunoglobulin, buffered aqueous solution
SAB1412631
ANTI-PRDM1 antibody produced in mouse, clone 4A6, purified immunoglobulin, buffered aqueous solution
HPA030033
Anti-PRDM1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2104001
Anti-PRDM1 antibody produced in rabbit, affinity isolated antibody
13204
β-tri-Calcium phosphate, puriss. p.a., ≥98% β-phase basis (unsintered powder)
Ca3O8P2
49963
β-tri-Calcium phosphate, puriss. p.a., ≥98% β-phase basis (sintered Powder)
Ca3O8P2
16483
β-tri-Calcium phosphate, puriss. p.a., ≥95% β-phase basis (sintered powder)
Ca3O8P2
21218
Calcium phosphate, purum p.a., ≥96.0% (calc. as Ca3(PO4)2, KT)
Ca3O8P2
I7034 Monoclonal Anti-Interleukin-4 antibody produced in mouse, clone 3007, purified immunoglobulin, lyophilized powder
I7159 Monoclonal Anti-Interleukin-4 antibody produced in mouse, clone 3010, purified immunoglobulin, lyophilized powder
WH0000639M1
Monoclonal Anti-PRDM1 antibody produced in mouse, clone 2B10, purified immunoglobulin, buffered aqueous solution
SAB5300402 Monoclonal Anti-PRDM1 antibody produced in mouse, clone 5E7, ascites fluid
SAB1403606
Monoclonal Anti-PRDM1, (N-terminal) antibody produced in mouse, clone 2F8, purified immunoglobulin, buffered aqueous solution