EMAIL THIS PAGE TO A FRIEND
Neuroscience letters
Impact of indomethacin on neuroinflammation and hippocampal neurogenesis in aged mice.
PMID 24796813
Abstract
Age-induced neuroinflammation could be a contributing factor to the restricted neurogenesis in aged mice. Indomethacin, a common non-steroidal anti-inflammatory drug, has been demonstrated to partially restore neurogenesis under pathophysiological inflammation-associated conditions in adult C57BL/6 mice. This study investigated whether indomethacin is able to decrease age-related neuroinflammation in the hippocampus (24-month-old mice) and thereby stimulate neurogenesis. During hippocampal aging, the transcript expression of pro-inflammatory cytokines (Tnfα, Il-1α, Il-1β), the chemokine Mip-1α, and markers for activated astrocytes (Gfap, Lcn2, but not Vim and Serpina3n) and microglia (Iba1, F4/80, Cd68, Cd86) significantly increased. Treatment with indomethacin significantly decreased COX-1 and COX-2 transcript expression. Of the age-related inflammatory mediators, only Gfap and Iba1 were affected by indomethacin treatment in the hippocampus, with a significantly reduced transcript expression being detected for both markers. Neurogenesis was unaffected by indomethacin. Thus, our data reveal that administration of indomethacin to aged mice is not able to effectively decrease neuroinflammation and promote neurogenesis.