EMAIL THIS PAGE TO A FRIEND
Foot and Ankle Surgery : Official Journal of the European Society of Foot and Ankle Surgeons
Successful arthroscopic decompression and synthetic grafting of a posterior talar cyst: a case report.
PMID 24796844