EMAIL THIS PAGE TO A FRIEND
Cardiovascular Toxicology
Low level tumor necrosis factor-alpha protects cardiomyocytes against high level tumor necrosis factor-alpha: brief insight into a beneficial paradox.
PMID 24798036