EMAIL THIS PAGE TO A FRIEND
Growth factors (Chur, Switzerland)
Inhibitory effects of lysophosphatidic acid receptor-5 on cellular functions of sarcoma cells.
PMID 24798396
Abstract
Lysophosphatidic acid (LPA) is a bioactive lipid that interacts with G protein-coupled LPA receptors (LPA receptor-1 (LPA1) to LPA6). Here, we investigated the effects of LPA signaling via LPA5 on cellular functions of sarcoma cells by generating Lpar5 overexpressing and Lpar5 knockdown cells from rat osteosarcoma and malignant fibrous histiocytoma cells, respectively. The cell motility activity of Lpar5 overexpressing cells was significantly lower, while Lpar5 knockdown cells showed high cell motility, compared with respective controls. Gelatin zymography showed that LPA5 suppressed the activation of matrix metalloproteinase-2. LPA5 also inhibited the cell motility activity of endothelial cells, correlating with the expression levels of vascular endothelial growth factor genes. These results suggest that LPA signaling via LPA5 negatively regulates the cellular functions of rat sarcoma cells.