EMAIL THIS PAGE TO A FRIEND
Journal of acquired immune deficiency syndromes (1999)
Bioequivalence of a dolutegravir, abacavir, and lamivudine fixed-dose combination tablet and the effect of food.
PMID 24798770
Abstract
The integrase inhibitor dolutegravir and nucleoside analogues abacavir and lamivudine are once-daily treatment options for HIV. This study (NCT01622790) evaluated, first, the bioequivalence (BE) of a fixed-dose combination (FDC) tablet containing dolutegravir 50 mg, abacavir 600 mg, and lamivudine 300 mg (dolutegravir/abacavir/lamivudine FDC) vs coadministered dolutegravir 50 mg and abacavir/lamivudine combination tablets (Epzicom) and, second, the effect of food on the dolutegravir/abacavir/lamivudine FDC tablet. Study part A (66 healthy subjects) was a single-dose, open-label, randomized, 2-period crossover study to evaluate the BE of the dolutegravir/abacavir/lamivudine FDC tablet and dolutegravir + abacavir/lamivudine tablets in the fasted state. In study part B, 12 subjects from part A received the dolutegravir/abacavir/lamivudine FDC tablet with a high-fat meal. BE and food effect were assessed by analysis of variance to determine the ratio of geometric least squares means and associated 90% confidence intervals for key pharmacokinetic parameters for each of dolutegravir, abacavir, and lamivudine. Sixty-two subjects completed part A. The dolutegravir/abacavir/lamivudine tablet was bioequivalent to the dolutegravir + abacavir/lamivudine tablets; 90% confidence intervals for the geometric least squares mean ratios fell within the 0.8-1.25 BE criteria. The effect of food on the dolutegravir/abacavir/lamivudine FDC tablet was similar to previous food effects observed with the separate formulations. The safety profile was comparable between treatments, with no observed serious or grade 3/4 adverse events. The BE of the dolutegravir/abacavir/lamivudine FDC tablet was demonstrated; it may be administered without regard to meals.