EMAIL THIS PAGE TO A FRIEND
Molecular cancer therapeutics
Inhibition of SIRT1 signaling sensitizes the antitumor activity of silybin against human lung adenocarcinoma cells in vitro and in vivo.
PMID 24798868
Abstract
Although silybin, a natural flavonolignan, has been shown to exhibit potent antitumor activities against various types of cancers, including lung cancer, the molecular mechanisms behind these activities remain unclear. Silent information regulator 1 (SIRT1) is a conserved NAD(+)-dependent deacetylase that has been implicated in the modulation of transcriptional silencing and cell survival. Furthermore, it plays a key role in carcinogenesis through the deacetylation of important regulatory proteins, including p53. In this study, we investigated the antitumor activity of silybin towards human lung adenocarcinoma cells in vitro and in vivo and explored the role of the SIRT1 signaling pathway in this process. Silybin treatment resulted in a dose- and time-dependent decrease in lung adenocarcinoma A549 cell viability. In addition, silybin exhibited strong antitumor activity illustrated by reductions in tumor cell adhesion, migratory capability, and glutathione levels and by increased apoptotic indices and reactive oxygen species levels. Silybin treatment also downregulated SIRT1 and upregulated p53 acetylation. SIRT1 siRNA (in vitro) or cambinol (a known SIRT1 inhibitor used for in vivo studies) further enhanced the antitumor activity of silybin. In summary, silybin is a potent inhibitor of lung adenocarcinoma cell growth that interferes with SIRT1 signaling, and this inhibition is a novel mechanism of silybin action that may be used for therapeutic intervention in lung adenocarcinoma treatment.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
EMU000411 MISSION® esiRNA, esiRNA targeting mouse Sirt1 (esiRNA1)