EMAIL THIS PAGE TO A FRIEND
Food Chemistry
Flavonol glycosides and other phenolic compounds in buds and leaves of different varieties of black currant (Ribes nigrum L.) and changes during growing season.
PMID 24799225