EMAIL THIS PAGE TO A FRIEND
Eksperimental'naia i klinicheskaia farmakologiia
[Calcium-depended mechanism of the antiplatelet activity of benzimidazole derivative RU-891].
PMID 24800520
Abstract
We have studied the effect of the new benzimidazole derivative RU-891, known to exhibit antiaggregant activity in vitro and in vivo, on the level of intracellular calcium ions in platelets of laboratory rabbits. Compound RU-891 was found to inhibit the thrombin-induced growth of intracellular calcium ion level in thrombocytes.This effect exceeded t he action o f reference d rugs and was not connected w ith the influence of RU-891 o n the potential-dependent c alci u m channels.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T4648 Thrombin from bovine plasma, lyophilized powder, 40-300 NIH units/mg protein (biuret)
T7513 Thrombin from bovine plasma, lyophilized powder, ≥2,000 NIH units/mg protein (E1%/280 = 19.5)
T6634 Thrombin from bovine plasma, lyophilized powder, 600-2,000 NIH units/mg protein (biuret)
T9549 Thrombin from bovine plasma, powder, cell culture tested, ≥1,500 NIH units/mg protein (E1%/280 = 19.5)
T7201 Thrombin from bovine plasma, lyophilized powder, ≥2,000 NIH units/mg protein (E1%/280 = 19.5)
T7326 Thrombin from bovine plasma, ≥60 NIH units/mg protein (biuret)
T6884
Thrombin from human plasma, lyophilized powder, ≥2,000 NIH units/mg protein (E1%/280, 18.3)
T7009
Thrombin from human plasma, lyophilized powder, ≥1,000 NIH units/mg protein (E1%/280, 18.3)
T1063
Thrombin from human plasma, lyophilized powder, ≥2800 NIH units/mg protein (E1%/280, 18.3)
T8885
Thrombin from human plasma, lyophilized powder, Suitable for routine use in the thrombin time test
T4393
Thrombin from human plasma, lyophilized powder, 1500-3500 NIH units/mg protein (E1%/280, 18.3), cell culture tested
T7572
Thrombin from human plasma, 400-1000 NIH units/mg protein
T5772 Thrombin from rat plasma, lyophilized powder, ≥1,000 units/mg protein (biuret)
T9326
Thrombin human, BioUltra, recombinant, expressed in HEK 293 cells, aqueous solution, ≥95% (SDS-PAGE)