PLoS ONE
Liver as a source for thymidine phosphorylase replacement in mitochondrial neurogastrointestinal encephalomyopathy.
PMID 24802030
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C3674
Citric acid trisodium salt, anhydrous, ≥98% (GC)
C6H5Na3O7