EMAIL THIS PAGE TO A FRIEND
The American journal of cardiology
Impact of albuminuria on the incidence of periprocedural myocardial injury in patients undergoing elective coronary stent implantation.
PMID 24802297
Abstract
Albuminuria has traditionally been associated with an elevated risk of cardiovascular events. However, few studies have examined the potential relation between albuminuria and periprocedural risk in percutaneous coronary intervention (PCI). The aim of this study was to evaluate the impact of albuminuria on the incidence of periprocedural myocardial injury (PMI) in patients who underwent PCI. The study included 252 consecutive patients who underwent PCI. The incidence of PMI was significantly higher in patients with albuminuria than in those with normoalbuminuria (31.9% vs 43.3%, respectively, p = 0.014). Even after adjustment for confounders, the presence of albuminuria predicted PMI (odds ratio 2.07, 95% confidence interval 1.08 to 3.97, p = 0.029). Furthermore, patients with albuminuria and preserved estimated glomerular filtration rate had a 4.2-fold higher risk for PMI than did patients with normoalbuminuria and preserved estimated glomerular filtration rate. In conclusion, albuminuria was a strong predictor of PMI in patients who underwent PCI.