EMAIL THIS PAGE TO A FRIEND
Journal of biomedical nanotechnology
Influence of silver nanoparticles on osteogenic differentiation of human mesenchymal stem cells.
PMID 24804548
Abstract
Silver nanoparticles (Ag NPs) have been widely used in bone grafts due to their antimicrobial/antiviral properties. Despite extensive research focused on the cytotoxicity and genotoxicity of Ag NPs, little is known of their effect on cell differentiation. In this study, the uptake of Ag NPs and its effect on the osteogenic differentiation of human mensenchymal stem cells (hMSCs) were investigated in vitro. Ag NPs with a diameter of -30 nm were prepared. The hMSCs were exposed to different concentrations of Ag NPs, i.e., 0, 10, 50 and 100 microg/mL. The results show that Ag NPs cause cytotoxicity of hMSCs. It is also demonstrated that the uptake of Ag NPs does not affect the alkaline phosphatase activity, osteocalcin gene expression, osteopontin expression and mineralization level of hMSCs. The uptake of Ag NPs does not influence the osteogenic differentiation of hMSCs. This will give a positive reference for the application of Ag NPs in bone tissue engineering, especially their incorporation with stem cells in bone grafts.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
85131
Silver, colloidal, 65-75% Ag basis
Ag
373249
Silver, evaporation slug, diam. × L 0.6 cm × 1.2 cm, 99.99% trace metals basis
Ag
327077
Silver, flakes, 10 μm, ≥99.9% trace metals basis
Ag
326976
Silver, foil, thickness 2.0 mm, 99.9% trace metals basis
Ag
348724
Silver, foil, thickness 1.5 mm, 99.9% trace metals basis
Ag
369438
Silver, foil, thickness 1.0 mm, 99.99% trace metals basis
Ag
265543
Silver, foil, thickness 1.0 mm, 99.9% trace metals basis
Ag
265535
Silver, foil, thickness 0.5 mm, 99.99% trace metals basis
Ag
345075
Silver, foil, thickness 0.5 mm, 99.9% trace metals basis
Ag
348716
Silver, foil, thickness 0.25 mm, 99.99% trace metals basis
Ag
326984
Silver, foil, thickness 0.25 mm, 99.9% trace metals basis
Ag
265527
Silver, foil, thickness 0.1 mm, 99.9% trace metals basis
Ag
326992
Silver, foil, thickness 0.075 mm, 99.9% trace metals basis
Ag
265519
Silver, foil, thickness 0.025 mm, 99.9% trace metals basis
Ag
303372
Silver, granular, >250 μm, 99.99% trace metals basis
Ag
461032
Silver, needles, 250-600 μm, ≥99.99% trace metals basis
Ag
265500
Silver, powder, <250 μm, 99.99% trace metals basis
Ag
327107
Silver, powder, <45 μm, ≥99.99% trace metals basis
Ag
327093
Silver, powder, 5-8 μm, ≥99.9% trace metals basis
Ag
327085
Silver, powder, 2-3.5 μm, ≥99.9% trace metals basis
Ag
484059
Silver, nanopowder, <150 nm particle size, 99% trace metals basis
Ag
265624
Silver, rod, diam. 7.0 mm, 99.99% trace metals basis
Ag
348775
Silver, rod, diam. 3.2 mm, 99.98% trace metals basis
Ag
204366
Silver, shot, 1-3 mm, 99.999% trace metals basis
Ag
327050
Silver, shot, 1-3 mm, ≥99.99% trace metals basis
Ag
267430
Silver, wire, diam. 2.0 mm, ≥99.99% trace metals basis
Ag
327018
Silver, wire, diam. 2.0 mm, 99.9% trace metals basis
Ag
348759
Silver, wire, diam. 1.5 mm, ≥99.99% trace metals basis
Ag
265594
Silver, wire, diam. 1.0 mm, ≥99.99% trace metals basis
Ag
265608
Silver, wire, diam. 1.0 mm, 99.9% trace metals basis
Ag