EMAIL THIS PAGE TO A FRIEND
The New England journal of medicine
Case records of the Massachusetts General Hospital. Case 14-2014. An 11-month-old girl with developmental delay.
PMID 24806163
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G8397 Monosialoganglioside GM2 from bovine brain, ≥95% (TLC)