EMAIL THIS PAGE TO A FRIEND
Biochimica et biophysica acta
Heteroglucan-dendrimer glycoconjugate: a modulated construct with augmented immune responses and signaling phenomena.
PMID 24806609
Abstract
Newer strategies for augmenting immune responses of pharmacologically active glucans may serve to improve the medicinal potential of these biomolecules. With this aim, the present work was focused on generating targeted high molecular size glucan particles with magnified immune response activity. Heteroglucans were conjugated with PAMAM dendrimers using a Schiff base reductive amination reaction to generate a polytethered molecule with multiple glucan motifs. The modulated construct was characterized by FTIR, TEM, (1)H NMR and dynamic light scattering (DLS) methods. Effects of conjugated glucans were examined in RAW 264.7 macrophage cells as well as in S-180 murine tumor models. Dendrimer-conjugated glucans were found to exhibit a two-fold increase in immune stimulation in comparison to unconjugated glucans. This may be corroborated by the predominant enhancement in immunological functions such as nitric oxide production, ROS generation and immune directed tumor inhibition in murine models. Immune cell surface markers (CD4, CD8, CD19, MHC-II) and cytokine levels were also found to be highly up-regulated in the splenocytes of mice subjected to particulate glucan administration. Our study also demonstrated that conjugated glucan treatment to RAW 264.7 cells strongly enhanced the phosphorylation of two downstream signalling molecules of the mitogen activated protein kinase (MAPKs) family: p38 and MEK1/2 relative to single glucans thereby relating molecular mechanisms with enhanced immune stimulation. The results obtained thus support that particulate format of soluble heteroglucan will thereby improve its functionality and identify leads in therapeutic competence.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M7556 Anti-MAP Kinase 2 (ERK-2) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution, ~1 mg/mL
HPA001633
Anti-MAP2K7 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2104449
Anti-MAP2K7 antibody produced in rabbit, affinity isolated antibody
SAB4502414
Anti-MAP2K7 antibody produced in rabbit, affinity isolated antibody
SAB1411096
Anti-MAP2K7 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB4300499
Anti-MAPK14 (Ab-182) antibody produced in rabbit, affinity isolated antibody
SAB1405665
Anti-MAPK14 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
AV06004
Anti-MAPK14 antibody produced in rabbit, affinity isolated antibody
M0800
Anti-p38 MAP Kinase antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB4500491
Anti-p38 MAPK antibody produced in rabbit, affinity isolated antibody
SAB4500492
Anti-p38 MAPK antibody produced in rabbit, affinity isolated antibody
SAB4500493
Anti-p38 MAPK antibody produced in rabbit, affinity isolated antibody
SAB4504623
Anti-phospho-MAP2K7 (pSer271) antibody produced in rabbit, affinity isolated antibody
SAB4504624
Anti-phospho-MAP2K7 (pThr275) antibody produced in rabbit, affinity isolated antibody
GW21175B Anti-RK antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB4502906
Anti-Synaptophysin antibody produced in rabbit, affinity isolated antibody
SAB4200544
Anti-Synaptophysin antibody, Mouse monoclonal, clone SVP-38, purified from hybridoma cell culture
HPA002858
Anti-SYP antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2102352
Anti-SYP antibody produced in rabbit, affinity isolated antibody
M7431 Monoclonal Anti-MAP Kinase 2 (ERK-2) antibody produced in mouse, clone 1B3B9, purified immunoglobulin, buffered aqueous solution
WH0005609M1
Monoclonal Anti-MAP2K7 antibody produced in mouse, clone 4E8, purified immunoglobulin, buffered aqueous solution
SAB1409528
Monoclonal Anti-MAP2K7 antibody produced in mouse, clone 6A4, purified immunoglobulin, buffered aqueous solution
SAB1409529
Monoclonal Anti-MAP2K7 antibody produced in mouse, clone 2C12, purified immunoglobulin, buffered aqueous solution
SAB1404255
Monoclonal Anti-MAP2K7, (N-terminal) antibody produced in mouse, clone 2G5, purified immunoglobulin, buffered aqueous solution
WH0006855M1
Monoclonal Anti-SYP antibody produced in mouse, clone 3B3, purified immunoglobulin, buffered aqueous solution
71435
Sodium cyanoborohydride, purum, ≥95.0% (RT)
NaBH3CN