EMAIL THIS PAGE TO A FRIEND
Drug metabolism and pharmacokinetics
Utility of cerebrospinal fluid drug concentration as a surrogate for unbound brain concentration in nonhuman primates.
PMID 24806821
Abstract
In central nervous system drug discovery, cerebrospinal fluid (CSF) drug concentration (C(CSF)) has been widely used as a surrogate for unbound brain concentrations (C(u,brain)). However, previous rodent studies demonstrated that when drugs undergo active efflux by transporters, such as P-glycoprotein (P-gp), at the blood-brain barrier, the C(CSF) overestimates the corresponding C(u,brain). To investigate the utility of C(CSF) as a surrogate for interstitial fluid (ISF) concentration (C(ISF)) in nonhuman primates, this study simultaneously determined the C(CSF) and C(ISF) of 12 compounds, including P-gp substrates, under steady-state conditions in cynomolgus monkeys using intracerebral microdialysis coupled with cisternal CSF sampling. Unbound plasma concentrations of non- or weak P-gp substrates were within 2.2-fold of the C(ISF) or C(CSF), whereas typical P-gp substrates (risperidone, verapamil, desloratadine, and quinidine) showed ISF-to-plasma unbound (K(p,uu,ISF)) and CSF-to-plasma unbound concentration ratios (K(p,uu,CSF)) that were appreciably lower than unity. Although the K(p,uu,CSF) of quinidine, verapamil, and desloratadine showed a trend of overestimating the K(p,uu,ISF), K(p,uu,CSF) showed a good agreement with K(p,uu,ISF) within 3-fold variations for all compounds examined. C(u,brain) of some basic compounds, as determined using brain homogenates, overestimated the C(ISF) and C(CSF). Therefore, C(CSF) could be used as a surrogate for C(ISF) in nonhuman primates.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P0884
(±)-Propranolol hydrochloride, ≥99% (TLC), powder
C16H21NO2 · HCl
40543
(±)-Propranolol hydrochloride, analytical reference material
C16H21NO2 · HCl
V4629
(±)-Verapamil hydrochloride, ≥99% (titration), powder
C27H38N2O4 · HCl
PHR1131
(±)-Verapamil hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
C27H38N2O4 · HCl
A5882
Antipyrine, analytical standard
C11H12N2O
10784
Antipyrine, tested according to Ph.Eur.
C11H12N2O
10790
Antipyrine, ≥99.0% (RT)
C11H12N2O
90567
Antipyrine, analytical reference material
C11H12N2O
C4024
Carbamazepine, powder
C15H12N2O
94496
Carbamazepine, analytical standard
C15H12N2O
C8981
Carbamazepine, meets USP testing specifications
C15H12N2O
C0450000
Carbamazepine, European Pharmacopoeia (EP) Reference Standard
C15H12N2O
1093001
Carbamazepine, United States Pharmacopeia (USP) Reference Standard
C15H12N2O
PHR1067
Carbamazepine, Pharmaceutical Secondary Standard; Certified Reference Material
C15H12N2O
D1069
Desloratadine, powder, ≥98% (HPLC)
C19H19ClN2
Y0001584
Desloratadine, European Pharmacopoeia (EP) Reference Standard
C19H19ClN2
1370280
Loratadine Related Compound A, United States Pharmacopeia (USP) Reference Standard
C19H19ClN2
1440808 Metoclopramide hydrochloride, United States Pharmacopeia (USP) Reference Standard
C14H22ClN3O2 · HCl · H2O
M0763
Metoclopramide hydrochloride, solid
C14H22ClN3O2 · HCl
Y0000197
Ondansetron for LC system suitability, European Pharmacopoeia (EP) Reference Standard
C18H19N3O · HCl · 2H2O
Y0000196
Ondansetron for TLC system suitability, European Pharmacopoeia (EP) Reference Standard
C18H19N3O · HCl · 2H2O
1478582
Ondansetron hydrochloride, United States Pharmacopeia (USP) Reference Standard
C18H19N3O · HCl · 2H2O
PHR1141
Ondansetron Hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
C18H19N3O · HCl · 2H2O
O3639
Ondansetron hydrochloride dihydrate, ≥98% (HPLC), powder
C18H19N3O · HCl · 2H2O
Y0000218
Ondansetron hydrochloride dihydrate, European Pharmacopoeia (EP) Reference Standard
C18H19N3O · HCl · 2H2O
1576005
Propranolol hydrochloride, United States Pharmacopeia (USP) Reference Standard
C16H21NO2 · HCl
P3500000
Propranolol hydrochloride, European Pharmacopoeia (EP) Reference Standard
C16H21NO2 · HCl
PHR1308
Propranolol hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
C16H21NO2 · HCl
P3600000
Propranolol hydrochloride for performance test, European Pharmacopoeia (EP) Reference Standard
C16H21NO2 · HCl
Q3625
Quinidine, anhydrous
C20H24N2O2