EMAIL THIS PAGE TO A FRIEND
Genes and Immunity
Contribution of increased ISG15, ISGylation and deregulated type I IFN signaling in Usp18 mutant mice during the course of bacterial infections.
PMID 24807690