EMAIL THIS PAGE TO A FRIEND
The American journal of tropical medicine and hygiene
A 41-year-old woman with migratory panniculitis.
PMID 24808244
Abstract
A 41-year-old woman had two months of intermittent migratory swellings in the trunk, face, and limbs associated with erythema, pruritus, and pain. Laboratory analysis showed moderate eosinophilia. The triad of eosinophilia, migratory lesions (nodular panniculitis), and raw fish consumption was highly suggestive of cutaneous gnathostomiasis. She was successfully treated with albendazole (400 mg twice a day for 21 days) and showed complete and permanent resolution of the lesions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
1012553
Albendazole, United States Pharmacopeia (USP) Reference Standard
C12H15N3O2S