EMAIL THIS PAGE TO A FRIEND
Fiziolohichnyi zhurnal (Kiev, Ukraine : 1994)
[The use of aluminum and its compounds for the biomedical purposes].
PMID 24809180
Abstract
Taking into consideration the nanodimension of functional components of living cells, the application of nanotechnology in the biomedical purposes is currently an urgent task. One of these directions is the use of aluminum nanoparticles for molecular diagnostics, targeted delivery of drugs, the development of new pharmaceuticals. The action of aluminum chloride on the regulatory mechanisms of smooth and skeletal muscle contraction and also the effect of aluminum ions on the ATPase activity and superprecipitation of actomyosin of different types of muscles is discussed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
310360
Aluminum, ACS reagent, 99%, wire, wire diam. ~1.5 mm
Al
518573
Aluminum, granular, <1 mm, 99.7% trace metals basis
Al
266558
Aluminum, wire, diam. 1.0 mm, 99.999% trace metals basis
Al
433705
Aluminum, evaporation slug, diam. × L 6.3 mm × 6.3 mm, 99.999% trace metals basis
Al
326852
Aluminum, foil, thickness 0.25 mm, 99.999% trace metals basis
Al
338788
Aluminum, pellets, 3-12 mm, 99.99% trace metals basis
Al
11009
Aluminum, powder, ≥91% (complexometric)
Al
266523
Aluminum, ReagentPlus®, beads, 5-15 mm, 99.9% trace metals basis
Al
326941
Aluminum, pellets, 3-8 mesh, ≥99.999% trace metals basis
Al
266574
Aluminum, foil, thickness 0.45-0.55 mm, 99.999% trace metals basis
Al
326860
Aluminum, foil, thickness 0.13 mm, ≥99.99% trace metals basis
Al
266515
Aluminum, ≤30 μm, spherical powder, ≥99%
Al
653608
Aluminum, powder, <5 μm particle size, 99.5% trace metals basis
Al
202584
Aluminum, powder, <75 μm, ≥99.95% trace metals basis
Al
266957
Aluminum, foil, thickness 1.0 mm, 99.999% trace metals basis
Al
733369
Aluminum, foil, thickness 8 μm, 99% trace metals basis
Al
326887
Aluminum, wire, diam. 0.58 mm, ≥99.99% trace metals basis
Al
202576
Aluminum, rod, diam. × L 3.0 mm × 100 mm, 99.999% trace metals basis
Al
GF26812043
Aluminum, foam, 150x150mm, thickness 3.2mm, bulk density 0.2g/cm3, porosity 0.93, 16 pores/cm, aluminum 6101 grade, 98.5%
Al
749036
Aluminum, sputtering target, diam. × thickness 3.00 in. × 0.125 in., 99.9995% trace metals basis
Al
GF33055185
Aluminum, foam, 150x150mm, thickness 6.35mm, bulk density 0.2g/cm3, porosity 0.93, 16 pores/cm, aluminum 6101 grade, 98.5%
Al
GF07499010
Aluminum, foam, 150x150mm, thickness 10mm, bulk density 0.2g/cm3, porosity 0.93, aluminum 6101 grade, 98.5%
Al
GF94706354
Aluminum, mesh, 150x150mm, nominal aperture 0.38mm, wire diameter 0.25mm, 40x40 wires/inch, open area 37%, plain weave mesh
Al
GF35817217
Aluminum, sphere, 200pcs, diameter 3.0mm, aluminum alloy 2017 precision sphere, grade 200, 95.2%
Al
GF37288020
Aluminum, rod, 1000mm, diameter 6.0mm, as drawn, 99.999%
Al
GF81923238
Aluminum, wire reel, 25m, diameter 0.25mm, as drawn, 99.5%
Al
GF98718935
Aluminum, sphere, 100pcs, diameter 1.0mm, aluminum alloy 2017 precision sphere, grade 200, 95.2%
Al
GF86715029
Aluminum, wire reel, 10m, diameter 0.015mm, hard, 99.99%
Al
GF43921638
Aluminum, wire reel, 5m, diameter 1.5mm, as drawn, 99.999%
Al
GF10029596
Aluminum, foam, 150x150mm, thickness 6.35mm, bulk density 0.2g/cm3, porosity 0.93, 8 pores/cm, aluminum 6101 grade, 98.5%
Al