EMAIL THIS PAGE TO A FRIEND
Environmental science and pollution research international
Concentration and distribution of antibiotics in water-sediment system of Bosten Lake, Xinjiang.
PMID 24809500
Abstract
This study investigated the residues of four sulfonamides, four quinolones, and four tetracyclines in surface water as well as surficial sediment samples, of Bosten Lake, in Xinjiang, China. The results showed the presence of 10 out of the 12 selected antibiotics in both water and sediment. Lomefloxacin was not detected in any of the samples. Among the 12 antibiotics considered, ciprofloxacin, with median concentrations of 39.22 ng L(-1) in surface water and 76.51 μg kg(-1) in surficial sediment, was the dominant antibiotic in all samples. The sorption coefficient values presented higher sorption capacities of tetracycline and chlortetracycline than the other antibiotics. The cluster analysis revealed elevated levels of pollution in sampling sites 1, 2, and 3, which were situated in a nearby urban area and in the estuary of Kaidu River. This study demonstrates the necessity of regulating the use of antibiotics and improving the management and treatment of their release.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
33703
Ofloxacin, VETRANAL, analytical standard
C18H20FN3O4
1478108
Ofloxacin, United States Pharmacopeia (USP) Reference Standard
C18H20FN3O4
PHR1168 Ofloxacin, Pharmaceutical Secondary Standard; Certified Reference Material
C18H20FN3O4
51239
Oxytetra Selective Supplement, for microbiology
C22H24N2O9
1491004
Oxytetracycline, United States Pharmacopeia (USP) Reference Standard
C22H24N2O9 · 2H2O
PHR1537
Oxytetracycline, Pharmaceutical Secondary Standard; Certified Reference Material
C22H24N2O9 · 2H2O
S8627
Sulfacetamide, ≥98.0%
C8H10N2O3S
46770
Sulfacetamide, VETRANAL, analytical standard
C8H10N2O3S
S8626
Sulfadiazine, ≥99.0%
C10H10N4O2S
35033
Sulfadiazine, VETRANAL, analytical standard
C10H10N4O2S
S1800000
Sulfadiazine, European Pharmacopoeia (EP) Reference Standard
C10H10N4O2S
1625009
Sulfadiazine, United States Pharmacopeia (USP) Reference Standard
C10H10N4O2S
PHR1343
Sulfadiazine, Pharmaceutical Secondary Standard; Certified Reference Material
C10H10N4O2S
Y0001480
Sulfadiazine for identification of impurity F, European Pharmacopoeia (EP) Reference Standard
C10H10N4O2S
S1975000
Sulfafurazole, European Pharmacopoeia (EP) Reference Standard
C11H13N3O3S
S7507
Sulfamethoxazole, analytical standard
C10H11N3O3S
76177
Sulfamethoxazole, certified reference material, TraceCERT®
C10H11N3O3S
31737
Sulfamethoxazole, VETRANAL, analytical standard
C10H11N3O3S
PHR1126
Sulfamethoxazole, Pharmaceutical Secondary Standard; Certified Reference Material
C10H11N3O3S
1631001
Sulfamethoxazole, United States Pharmacopeia (USP) Reference Standard
C10H11N3O3S
S2100000
Sulfamethoxazole, European Pharmacopoeia (EP) Reference Standard
C10H11N3O3S
31739
Sulfisoxazole, VETRANAL, analytical standard
C11H13N3O3S