PloS one
Changes in the expression of the Toll-like receptor system in the aging rat kidneys.
PMID 24810370
Abstract
The mechanisms of kidney aging are not yet clear. Studies have shown that immunological inflammation is related to kidney aging. Toll-like receptors (TLRs) are one of the receptor types of the body's innate immune system. The function of the TLR system and the mechanisms by which it functions in renal aging remain unclear. In the present study, we, for the first time, systematically investigated the role of the TLR system and the inflammation responses activated by TLRs during kidney aging. We used western blot and immunohistochemistry to systematically analyze the changes in the expression and activation of the endogenous TLR ligands HSP70 and HMGB1, the TLRs (TLR1-TLR11), their downstream signaling pathway molecules MyD88 and Phospho-IRF-3, and the NF-κB signaling pathway molecules Phospho-IKKβ, Phospho-IκBα (NF-κB inhibition factor α), NF-κBp65, and Phospho-NF-κBp65 (activated NF-κB p65) in the kidneys of 3 months old (youth group), 12 months old (middle age group), and 24 months old (elderly group) rats. We used RT-qPCR to detect the mRNA expression changes of the proinflammatory cytokines CCL3, CCL4, CCL5, CD80, TNF-α, and IL-12b in the rat renal tissues of the various age groups. We found that during kidney aging, the HSP70 and HMGB1 expression levels were significantly increased, and the expression levels of TLR1, 2, 3, 4, 5, and 11 and their downstream signaling pathway molecules MyD88 and Phospho-IRF-3 were markedly elevated. Further studies have shown that in the aging kidneys, the expression levels of the NF-κB signaling pathway molecules Phospho-IKKβ, Phospho-IκBα, NF-κBp65, and Phospho-NF-κBp65 were obviously increased, and those of the proinflammatory cytokines CCL3, CCL4, CCL5, CD80, TNF-α, and IL-12b were significantly upregulated. These results showed that the TLR system might play an important role during the kidney aging process maybe by activating the NF-κB signaling pathway and promoting the high expression of inflammation factors.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AV38110
Anti-HMGB1 (AB2) antibody produced in rabbit, IgG fraction of antiserum
SAB4501401
Anti-HMGB1 antibody produced in rabbit, affinity isolated antibody
SAB2101049
Anti-HMGB1 antibody produced in rabbit, affinity isolated antibody
SAB3500280
Anti-IRF3 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA004895
Anti-IRF3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV31992
Anti-IRF3 antibody produced in rabbit, affinity isolated antibody, lyophilized powder
PRS3639
Anti-TLR1 (ab1) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
PRS3641
Anti-TLR1 (ab2) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2900396
Anti-TLR1 (AB2) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
PRS3275
Anti-TLR10 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
PRS3135
Anti-TLR2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
PRS3643
Anti-TLR3 (ab1) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
PRS3645
Anti-TLR3 (ab2) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2900405
Anti-TLR3 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB1412548
ANTI-TLR4 antibody produced in mouse, clone 4B10, purified immunoglobulin, buffered aqueous solution
SAB1412602
ANTI-TLR4 antibody produced in mouse, clone 3G12, purified immunoglobulin, buffered aqueous solution
SAB3500374
Anti-TLR5 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
PRS3651
Anti-TLR6 (ab1) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
PRS3653
Anti-TLR6 (ab2) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA059613
Anti-TLR7 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
PRS3269
Anti-TLR7 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1412111 ANTI-TLR9 antibody produced in mouse, clone 2D11, purified immunoglobulin, buffered aqueous solution
SAB1412155
ANTI-TLR9 antibody produced in mouse, clone 3B7, purified immunoglobulin, buffered aqueous solution
SAB1412156
ANTI-TLR9 antibody produced in mouse, clone 2H5, purified immunoglobulin, buffered aqueous solution
SAB1412299
ANTI-TLR9 antibody produced in mouse, clone 1E4, purified immunoglobulin, buffered aqueous solution
SAB1412300
ANTI-TLR9 antibody produced in mouse, clone 2G3, purified immunoglobulin, buffered aqueous solution
SAB1412302
ANTI-TLR9 antibody produced in mouse, clone 2C3, purified immunoglobulin, buffered aqueous solution
SAB1404475
Monoclonal Anti-TLR4 antibody produced in mouse, clone 3B6, purified immunoglobulin, buffered aqueous solution
SAB1402375
Monoclonal Anti-TLR4 antibody produced in mouse, clone 1H7, purified immunoglobulin, buffered aqueous solution
WH0054106M3
Monoclonal Anti-TLR9 antibody produced in mouse, clone 1E8, purified immunoglobulin, buffered aqueous solution