EMAIL THIS PAGE TO A FRIEND
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine
Long noncoding RNA SPRY4-IT1 is upregulated in esophageal squamous cell carcinoma and associated with poor prognosis.
PMID 24810925
Abstract
LncRNA SPRY4-IT1 has been shown to promote the progression of melanoma. However, the role of lncRNA SPRY4-IT1 in human esophageal squamous cell carcinoma (ESCC) remains unclear. The purpose of this study is to investigate the clinical significance and biological functions of SPRY4-IT1 in ESCC. The expression levels of lncRNA SPRY4-IT in 92 ESCC patients and 8 ESCC cell lines were evaluated by quantitative reverse transcriptase polymerase chain reaction (qRT-PCR). The prognostic significance was evaluated using Kaplan-Meier and Cox regression analyses. Small interfering RNA (siRNA) was used to suppress SPRY4-IT1 expression in ESCC cell lines. Both in vitro and in vivo assays were performed to further explore its role in tumor progression. SPRY4-IT1 levels were significantly higher in ESCC tissues and cells than in corresponding adjacent noncancerous tissues and nontumorigenic esophageal epithelial cells, and the ESCC patients with higher SPRY4-IT1 expression had an advanced clinical stage and poorer prognosis than those with lower SPRY4-IT1 expression. The multivariate analysis revealed that SPRY4-IT1 expression level is an independent prognostic factor in ESCC patients. In vitro assays demonstrated that knockdown of SPRY4-IT1 reduced cell proliferation, invasiveness, and migration. In vivo assays demonstrated that knockdown of SPRY4-IT1 decreases cell growth. SPRY4-IT1 is a novel molecule involved in ESCC progression, which may provide a potential prognostic biomarker and a potential target for therapeutic intervention.