EMAIL THIS PAGE TO A FRIEND
Fish & Shellfish Immunology
Transgenic expression of salmon delta-5 and delta-6 desaturase in zebrafish muscle inhibits the growth of Vibrio alginolyticus and affects fish immunomodulatory activity.
PMID 24811009