EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the USA
Monoclonal antibodies against heparin-binding growth factor II/basic fibroblast growth factor that block its biological activity: invalidity of the antibodies for tumor angiogenesis.
PMID 2481318