EMAIL THIS PAGE TO A FRIEND
Molecular oncology
Caspase-8 mutations in head and neck cancer confer resistance to death receptor-mediated apoptosis and enhance migration, invasion, and tumor growth.
PMID 24816188
Abstract
Little is known regarding molecular markers in head and neck squamous cell carcinoma (HNSCC) that predict responsiveness to different therapeutic regimens or predict HNSCC progression. Mutations in procaspase-8 occur in 9% of HNSCC primary tumors, but the functional consequences of these mutations are poorly understood. In this study, we examined the impact of four, representative, HNSCC-associated procaspase-8 mutations on activation of the extrinsic apoptosis pathway, as well as cellular migration and invasion, and in vivo tumor growth. All four mutant proteins acted to potently inhibit activation of apoptosis following treatment with TRAIL or agonistic anti-Fas. In contrast to wild-type procaspase-8, the mutant proteins were not recruited to FADD following treatment with TRAIL or anti-Fas, but may be constitutively bound by FADD. Three of the four procaspase-8 mutants promoted enhanced cellular migration and invasion through matrigel, relative to that seen with the wild-type procaspase-8 protein. Procaspase-8 mutation also stimulated the growth of HNSCC xenograft tumors. These findings indicate that HNSCC-associated procaspase-8 mutations inhibit activation of the extrinsic apoptosis pathway and are likely to represent markers for resistance to therapeutic regimens incorporating death receptor activators. Moreover, procaspase-8 mutations may serve as markers of HNSCC tumor progression, as exemplified by enhanced migration, invasion, and tumor growth.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB2501669
Anti-CASP3 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA002643
Anti-CASP3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV00021
Anti-CASP3 antibody produced in rabbit, IgG fraction of antiserum
SAB4503291
Anti-CASP3 antibody produced in rabbit, affinity isolated antibody
HPA001302
Anti-CASP8 antibody produced in rabbit, Ab3, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA005688
Anti-CASP8 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
C9598 Anti-Caspase 3 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
GW21154 Anti-Caspase 8 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
C4106 Anti-Caspase 8 antibody, Mouse monoclonal, clone CAS8, purified from hybridoma cell culture
SAB3500404
Anti-Caspase-8 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
AV01012
Anti-FADD (AB1) antibody produced in rabbit, affinity isolated antibody
AV30294
Anti-FADD (AB2) antibody produced in rabbit, affinity isolated antibody
HPA001464
Anti-FADD antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4503471
Anti-FADD antibody produced in rabbit, affinity isolated antibody
F8053 Anti-FADD antibody, Mouse monoclonal, clone FD19, purified from hybridoma cell culture
GW22766 Anti-FAS (ab1) antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
GW22767 Anti-FAS (ab2) antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB2501317
Anti-FAS antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA027444
Anti-FAS antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2105218
Anti-FAS antibody produced in rabbit, affinity isolated antibody
SAB4500487
Anti-PARP (Cleaved-Asp214) antibody produced in rabbit, affinity isolated antibody
SAB4500488
Anti-PARP (Cleaved-Gly215) antibody produced in rabbit, affinity isolated antibody
P7605 Anti-PARP antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
AV33754
Anti-PARP1 antibody produced in rabbit, IgG fraction of antiserum
HPA045168
Anti-PARP1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
WH0000836M2
Monoclonal Anti-CASP3 antibody produced in mouse, clone 4D3, purified immunoglobulin, buffered aqueous solution
SAB5300353 Monoclonal Anti-CASP8 antibody produced in mouse, clone 1H11, ascites fluid
C5737 Monoclonal Anti-Caspase 3 antibody produced in mouse, clone 84803.111, purified immunoglobulin, lyophilized powder
SAB4700005 Monoclonal Anti-CD95 antibody produced in mouse, clone LT95, purified immunoglobulin, buffered aqueous solution
WH0008772M1
Monoclonal Anti-FADD antibody produced in mouse, clone 3A12, purified immunoglobulin, buffered aqueous solution