EMAIL THIS PAGE TO A FRIEND
Journal of Cell Science
RasGAP mediates neuronal survival in Drosophila through direct regulation of Rab5-dependent endocytosis.
PMID 24816559
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
61739
N-Lauroylsarcosine, purum p.a., ≥98.0% (GC)
C15H29NO3
L5000
N-Lauroylsarcosine, neat, ≥95%
C15H29NO3