EMAIL THIS PAGE TO A FRIEND
Journal of Pharmacology and Experimental Therapeutics
Potent inhibition of alcohol self-administration in alcohol-preferring rats by a ?-opioid receptor antagonist.
PMID 24817033