EMAIL THIS PAGE TO A FRIEND
Analytical and Bioanalytical Chemistry
A sensitive and accurate quantitative method to determine N-arachidonoyldopamine and N-oleoyldopamine in the mouse striatum using column-switching LC-MS-MS: use of a surrogate matrix to quantify endogenous compounds.
PMID 24817351
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A8848
Arachidonoyl dopamine, ≥98% (HPLC), ethanol solution
C28H41NO3