EMAIL THIS PAGE TO A FRIEND
Journal of ethnopharmacology
Paris saponin VII from trillium tschonoskii reverses multidrug resistance of adriamycin-resistant MCF-7/ADR cells via P-glycoprotein inhibition and apoptosis augmentation.
PMID 24818584
Abstract
Saponins of several herbs are known to induce apoptosis in some cancer cells and are proposed to be promising modulators of drug resistance. In the present study, we extracted Paris saponin VII (PS VII), a kind of saponin, from Trillium tschonoskii Maxim. and observed its effect on adriamycin-resistant breast cancer cells. An adriamycin-resistant human breast cancer cell line, MCF-7/ADR cells were exposed to different concentrations of PS VII (0-100 μmol/L). Then, flow cytometric assays and a human apoptosis array were used to detect apoptotic cells and apoptosis related protein expression. P-glycoprotein levels and intracellular rhodamine 123 (RH-123) accumulations were measured to evaluate the expression and activity of P-glycoprotein. PS VII dose dependently suppressed cell viability as well as triggered apoptosis and modulated drug resistance of MCF-7/ADR cells. Further results showed that PS VII treatment in MCF-7/ADR cells led to increased TNFR1, TRAIL R1/DR4, TRAIL R2/DR5, and FADD expression, and activation of PARP, caspase-8, and 3. In parallel to the alterations, P-glycoprotein expression and activity were also reduced. These findings showed that PS VII might be an effective tumouristatic agent for the treatment of MDR breast cancer.