PloS one
The Drosophila Su(var)3-7 gene is required for oogenesis and female fertility, genetically interacts with piwi and aubergine, but impacts only weakly transposon silencing.
PMID 24820312
Abstract
Heterochromatin is made of repetitive sequences, mainly transposable elements (TEs), the regulation of which is critical for genome stability. We have analyzed the role of the heterochromatin-associated Su(var)3-7 protein in Drosophila ovaries. We present evidences that Su(var)3-7 is required for correct oogenesis and female fertility. It accumulates in heterochromatic domains of ovarian germline and somatic cells nuclei, where it co-localizes with HP1. Homozygous mutant females display ovaries with frequent degenerating egg-chambers. Absence of Su(var)3-7 in embryos leads to defects in meiosis and first mitotic divisions due to chromatin fragmentation or chromosome loss, showing that Su(var)3-7 is required for genome integrity. Females homozygous for Su(var)3-7 mutations strongly impair repression of P-transposable element induced gonadal dysgenesis but have minor effects on other TEs. Su(var)3-7 mutations reduce piRNA cluster transcription and slightly impact ovarian piRNA production. However, this modest piRNA reduction does not correlate with transposon de-silencing, suggesting that the moderate effect of Su(var)3-7 on some TE repression is not linked to piRNA production. Strikingly, Su(var)3-7 genetically interacts with the piwi and aubergine genes, key components of the piRNA pathway, by strongly impacting female fertility without impairing transposon silencing. These results lead us to propose that the interaction between Su(var)3-7 and piwi or aubergine controls important developmental processes independently of transposon silencing.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
06-911
Anti-acetyl-Histone H3 (Lys14) Antibody, Use Anti-acetyl-Histone H3 (Lys14) Antibody (Rabbit Polyclonal Antibody) validated in ICC, IP, WB to detect acetyl-Histone H3 (Lys14) also known as H3K14Ac, Histone H3 (acetyl K14).
06-570
Anti-phospho-Histone H3 (Ser10) Antibody, Mitosis Marker, Anti-phospho-Histone H3 (Ser10) Antibody, Mitosis Marker is a Rabbit Polyclonal Antibody for detection of Histone H3 phosphorylated at serine 10.This highly published Ab, also known as Anti-H3S10p, has been validated in ICC, IP & WB.
07-442
Anti-trimethyl-Histone H3 (Lys9) Antibody, Use Anti-trimethyl-Histone H3 (Lys9) Antibody (rabbit polyclonal antibody) validated in DB, ICC, Mplex, PIA, WB, ChIP-seq to detect trimethyl-Histone H3 (Lys9) also known as H3K9me3, Histone H3 (tri methyl K9).
SAB1411733 Monoclonal Anti-HA antibody produced in mouse, clone 1B8, purified immunoglobulin, buffered aqueous solution
SAB1411734 Monoclonal Anti-HA antibody produced in mouse, clone 4D4, purified immunoglobulin, buffered aqueous solution
SAB1411735
Monoclonal Anti-HA antibody produced in mouse, clone 1F7, purified immunoglobulin, buffered aqueous solution
SAB1411737
Monoclonal Anti-HA antibody produced in mouse, clone 3C6, purified immunoglobulin, buffered aqueous solution
SAB1411738
Monoclonal Anti-HA antibody produced in mouse, clone 3F6, purified immunoglobulin, buffered aqueous solution