EMAIL THIS PAGE TO A FRIEND
Endocrinology
Anoctamin 1 calcium-activated chloride channel downregulates estrogen production in mouse ovarian granulosa cells.
PMID 24823391
Abstract
Calcium-dependent chloride conductances have been described in chicken and human granulosa cells (GCs) and may be involved in steroidogenesis. However, the molecular identities of corresponding chloride channels remain unknown. The purpose of this study was to explore the expression and function of the Anoctamin 1 (ANO1) calcium-activated chloride channel (CaCC) in mouse ovary. ANO1 mRNA and protein expression was identified in mouse ovary GCs by RT-PCR, immunoblot, and immunostaining. Patch-clamp analysis on freshly isolated GCs identified an outwardly rectifying Ca(2+)-activated Cl(-) current that was completely blocked by a selective ANO1 inhibitor T16Ainh-A01. Knockdown of ANO1 mRNA or incubation with a selective inhibitor T16Ainh-A01 enhanced estradiol production, whereas a selective ANO1 activator Eact significantly inhibited estradiol production in primary cultured GCs. The ANO1 expression or activation increases the phosphorylation of ERK1/2 and decreases aromatase expression. The ANO1 expression level is remarkably higher at the proestrous and estrous stages in the estrous cycle. In vivo study indicated a profound induction of ANO1 expression in ovarian GCs by pregnant mare serum gonadotropin (PMSG) that can be further augmented by hCG treatment, suggesting that both FSH and LH may upregulate ANO1 expression at the proestrous and estrous stages. ANO1 expression was remarkably reduced in DHEA-induced PCOS ovary. These data identified for the first time the expression of ANO1 Ca(2+) activated Cl(-) channel in mouse ovarian GCs and determined its negative regulation on estrogen production possibly through MEK-ERK signaling cascade. The present study provided new insights into the molecular mechanisms for the regulation of folliculogenesis and ovulation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
HPA032148
Anti-ANO1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA051194
Anti-CYP19A1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1410280
Anti-CYP19A1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB4500606
Anti-Cytochrome P450 19A1 antibody produced in rabbit, affinity isolated antibody
SAB1411096
Anti-MAP2K7 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB4502414
Anti-MAP2K7 antibody produced in rabbit, affinity isolated antibody
HPA001633
Anti-MAP2K7 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB3500318
Anti-TMEM16A antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2102460
Anti-TMEM16A antibody produced in rabbit, affinity isolated antibody
I3146 ITS Liquid Media Supplement (100×), liquid, sterile-filtered, BioReagent, suitable for cell culture
WH0001588M3
Monoclonal Anti-CYP19A1 antibody produced in mouse, clone 2B6, purified immunoglobulin, buffered aqueous solution
SAB1409528
Monoclonal Anti-MAP2K7 antibody produced in mouse, clone 6A4, purified immunoglobulin, buffered aqueous solution
SAB1409529
Monoclonal Anti-MAP2K7 antibody produced in mouse, clone 2C12, purified immunoglobulin, buffered aqueous solution
WH0005609M1
Monoclonal Anti-MAP2K7 antibody produced in mouse, clone 4E8, purified immunoglobulin, buffered aqueous solution
SAB1404255
Monoclonal Anti-MAP2K7, (N-terminal) antibody produced in mouse, clone 2G5, purified immunoglobulin, buffered aqueous solution
SAB2104449
Anti-MAP2K7 antibody produced in rabbit, affinity isolated antibody