EMAIL THIS PAGE TO A FRIEND
Investigative ophthalmology & visual science
Expression of vascular endothelial growth factor in human ocular adnexal lymphoma.
PMID 24825110
Abstract
To examine the expression of VEGF in extranodal marginal zone B-cell lymphoma (EMZL) and reactive lymphoid hyperplasia (RLH) of human ocular adnexa, and analyze the correlation with the intratumoral microvessel density (MVD). Twenty-two EMZL and 16 RLH tissues were examined in this study. Paraformaldehyde-fixed, paraffin-embedded tissue sections were processed for immunohistochemistry with antibodies against VEGF and CD20. Vascular endothelial growth factor expression was analyzed using the ELISA and RT-PCR in the EMZL tissues. Microvessel density was determined based on the immunoreactivity for anti-CD34 antibody. Vascular endothelial growth factor immunoreactivity was detected in the cytoplasm of lymphoid cells in EMZL and RLH. ELISA and RT-PCR confirmed VEGF protein and mRNA expressions in the EMZL tissue, respectively. Vascular endothelial growth factor-immunopositive rate in B-cells was significantly higher in 12 conjunctival EMZLs than four RLHs (P < 0.01) and 10 orbital EMZLs than 12 RLHs (P < 0.05). The MVD showed a significant positive correlation with the VEGF-immunopositive rate in conjunctival and orbital EMZLs. This study demonstrated increased VEGF expression in human conjunctival and orbital EMZL compared with that in RLH, suggesting that VEGF plays a significant role in the pathogenesis and tumor angiogenesis of ocular adnexal lymphoma.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1405580
Anti-MS4A1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA014341
Anti-MS4A1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA014391
Anti-MS4A1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
RAB0508
Human VEGF ELISA Kit, for cell and tissue lysates
RAB0328
Human VEGF R2 ELISA Kit, for serum, plasma, cell culture supernatant and urine
RAB0195
Human VEGF R3 ELISA Kit, for serum, plasma, cell culture supernatant, urine
RAB0507
Human VEGF-A ELISA Kit, for serum, plasma, cell culture supernatants and urine
SAB4700114 Monoclonal Anti-CD20 antibody produced in mouse, clone 2H7, purified immunoglobulin, buffered aqueous solution
SAB4700119 Monoclonal Anti-CD20 antibody produced in mouse, clone MEM-269, purified immunoglobulin, buffered aqueous solution
SAB4700121
Monoclonal Anti-CD20 antibody produced in mouse, clone MEM-97, purified immunoglobulin, buffered aqueous solution
SAB1403646
Monoclonal Anti-MS4A1 antibody produced in mouse, clone 4F11, purified immunoglobulin, buffered aqueous solution
WH0000931M1
Monoclonal Anti-MS4A1 antibody produced in mouse, clone 5C11, purified immunoglobulin, buffered aqueous solution
RAB0509
Mouse VEGF ELISA Kit, for serum, plasma, cell culture supernatant
RAB0510
Mouse VEGF ELISA Kit, for cell and tissue lysates
RAB0511
Rat VEGF ELISA Kit, for serum, plasma and cell culture supernatant
RAB0512
Rat VEGF ELISA Kit, for cell and tissue lysates