EMAIL THIS PAGE TO A FRIEND
Journal of clinical pathology
ERBB3 as an independent prognostic marker for nasopharyngeal carcinoma.
PMID 24825912
Abstract
Although the ERBB proteins have been shown to be associated in many types of human tumours and serve as important cancer therapeutic targets, however, data regarding the expression and clinical relevance of ERBBs in nasopharyngeal carcinoma (NPC) are still conflicting. The aim of this study is to investigate the expression pattern of all ERBB members simultaneously in NPC tissues using immunohistochemistry and determine their clinical relevance. The expression of all members of ERBB proteins was evaluated using immunohistochemistry on 82 NPC tissue samples. Relationship between the ERBB protein expression, clinicopathological parameters and patient outcome was assessed using univariate and multivariate analyses. We found that ERBB1, ERBB2 and ERBB3 were strongly expressed in the normal nasopharyngeal epithelial cells. A marked reduction of ERBB1 and ERBB2 expression in NPC was observed compared with the non-cancerous tissues. 76 of 82 (92.7%) cases were ERBB3-positive, while ERBB4 was not expressed in both normal and NPC. The univariate log-rank analysis showed that regional lymph node metastasis, systemic metastasis, recurrence and ERBB3 expression were associated with patient survival. The ERBB3 expression was not correlated to other clinicopathological factors. Furthermore, multivariate analysis revealed that ERBB3 expression was an independent prognostic factor influencing patient survival. Our results suggested that the expression of ERBB3 is associated with patient survival and could serve as a novel and valuable predictor for prognostic evaluation of patients with NPC.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4300352
Anti-EGFR (Ab-1070) antibody produced in rabbit, affinity isolated antibody
SAB4300353
Anti-EGFR (Ab-1092) antibody produced in rabbit, affinity isolated antibody
SAB4300510
Anti-EGFR (Ab-1110) antibody produced in rabbit, affinity isolated antibody
SAB4300472
Anti-EGFR (Ab-1172) antibody produced in rabbit, affinity isolated antibody
SAB4300480
Anti-EGFR (Ab-1197) antibody produced in rabbit, affinity isolated antibody
SAB4300453
Anti-EGFR (Ab-678) antibody produced in rabbit, affinity isolated antibody
SAB4300454
Anti-EGFR (Ab-693) antibody produced in rabbit, affinity isolated antibody
SAB4300481
Anti-EGFR (Ab-869) antibody produced in rabbit, affinity isolated antibody
HPA001200
Anti-EGFR antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA018530
Anti-EGFR antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4500711
Anti-EGFR antibody produced in rabbit, affinity isolated antibody
SAB4500712
Anti-EGFR antibody produced in rabbit, affinity isolated antibody
SAB4500720
Anti-EGFR antibody produced in rabbit, affinity isolated antibody
PLA0101
Rabbit anti-EGFR Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.