EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the USA
Polarized deposition of basement membrane proteins depends on Phosphatidylinositol synthase and the levels of Phosphatidylinositol 4,5-bisphosphate.
PMID 24828534