EMAIL THIS PAGE TO A FRIEND
Journal of cell science
Intracellular localization of the M1 muscarinic acetylcholine receptor through clathrin-dependent constitutive internalization is mediated by a C-terminal tryptophan-based motif.
PMID 24829147
Abstract
The M1 muscarinic acetylcholine receptor (M1-mAChR, encoded by CHRM1) is a G-protein-coupled membrane receptor that is activated by extracellular cholinergic stimuli. Recent investigations have revealed the intracellular localization of M1-mAChR. In this study, we observed constitutive internalization of M1-mAChR in mouse neuroblastoma N1E-115 cells without agonist stimulation. Constitutive internalization depended on dynamin, clathrin and the adaptor protein-2 (AP-2) complex. A WxxI motif in the M1-mAChR C-terminus is essential for its constitutive internalization, given that replacement of W(442) or I(445) with alanine residues abolished constitutive internalization. This WxxI motif resembles YxxΦ, which is the canonical binding motif for the μ2 subunit of the AP-2 complex. The M1-mAChR C-terminal WxxI motif interacted with AP-2 μ2. W442A and I445A mutants of the M1-mAChR C-terminal sequence lost AP-2-μ2-binding activity, whereas the W442Y mutant bound more effectively than wild type. Consistent with these results, W442A and I445A M1-mAChR mutants selectively localized to the cell surface. By contrast, the W442Y receptor mutant was found only at intracellular sites. Our data indicate that the cellular distribution of M1-mAChR is governed by the C-terminal tryptophan-based motif, which mediates constitutive internalization.