EMAIL THIS PAGE TO A FRIEND
Journal of the American Academy of Dermatology
Infection with Mycobacterium fortuitum during acupoint embedding therapy.
PMID 24831334
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
1134335
Ciprofloxacin hydrochloride, United States Pharmacopeia (USP) Reference Standard
C17H18FN3O3 · HCl · H2O