EMAIL THIS PAGE TO A FRIEND
Journal of Chromatographic Science
Development and validation of liquid chromatography-tandem mass spectrometric method for the quantification of ciprofibrate from human plasma.
PMID 24833347