EMAIL THIS PAGE TO A FRIEND
The Journal of Antimicrobial Chemotherapy
Activity of colistin combined with doripenem at clinically relevant concentrations against multidrug-resistant Pseudomonas aeruginosa in an in vitro dynamic biofilm model.
PMID 24833752
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML1220
Doripenem hydrate, ≥98% (HPLC)
C15H24N4O6S2 · H2O