EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the United States of America
Reply to Hristov et al.: Linking methane emissions inventories with atmospheric observations.
PMID 24834496