EMAIL THIS PAGE TO A FRIEND
Revue medicale suisse
[Rumbling, spasms and diarrhea: do not throw more!].
PMID 24834641
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G5004 Gluten from wheat