EMAIL THIS PAGE TO A FRIEND
European journal of cancer (Oxford, England : 1990)
Identification and characterisation of a novel heat shock protein 90 inhibitor ONO4140.
PMID 24835034
Abstract
Heat shock protein (Hsp) 90 is a key component of the super-chaperone complex that maintains functionally active conformation of various client proteins. Many of these client proteins regulate important nodal points in multiple signalling pathways that promote cancer cell growth and survival. Inhibitors of Hsp90, therefore, have the potential of functioning as anti-cancer agents with pleiotropic effects. Identification of novel Hsp90 inhibitors with more favourable pharmacological properties is a priority in cancer therapy. To achieve this goal, we screened a compound library using a biochemical assay based on refolding of denatured firefly luciferase. The assay revealed high sensitivity, reliability and reproducibility with a Z-factor of 0.81 ± 0.17. Six Hsp90 inhibitory compounds identified by this screening with IC50 values between 1.0 and 6 μM were further characterised for anti-proliferative activity by Cell Titer-Blue Cell Viability Assay using multiple tumour cell lines. Of particular interest was ONO4140 with lowest GI50 values in three different cancer cell lines viz; DU-145, BT-474 and K562 cell lines. This study also revealed that short-term exposure of tumour cells with ONO4140 is sufficient to inhibit the catalytic activity of Hsp90, evaluated through disruption of Hsp90-p23 association by immunoprecipitation. This short term exposure appears to initiate events like degradation of Hsp90 client proteins such as ErbB2/Her-2 and Akt with concomitant inhibition of survival signalling leading to the apoptotic death of tumour cells as seen by western blotting and Caspase Glow-3,7 assay. The study also reveals that apoptosis following Hsp90 inhibition with ONO4140 occurs via Caspase9-Caspase3 intrinsic apoptotic pathway, a process that is likely triggered by inactivation of Akt. In conclusion, we have identified a novel class of synthetic compounds which show potent Hsp90 inhibitory action in preclinical studies. The discovery of this novel class of synthetic Hsp90 inhibitors with simple chemical backbone allows us to conduct further structural modifications to improve their potency and pharmacokinetic properties for use in cancer therapy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4500487
Anti-PARP (Cleaved-Asp214) antibody produced in rabbit, affinity isolated antibody
SAB4500488
Anti-PARP (Cleaved-Gly215) antibody produced in rabbit, affinity isolated antibody
P7605 Anti-PARP antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
AV33754
Anti-PARP1 antibody produced in rabbit, IgG fraction of antiserum
HPA045168
Anti-PARP1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
WH0000835M10
Monoclonal Anti-CASP2 antibody produced in mouse, clone 1C11, purified immunoglobulin, buffered aqueous solution
WH0000842M1
Monoclonal Anti-CASP9 antibody produced in mouse, clone 3B8-4G2, purified immunoglobulin, buffered aqueous solution
SAB1403634
Monoclonal Anti-CASP9 antibody produced in mouse, clone 4F12-G6, purified immunoglobulin, buffered aqueous solution
C4356
Monoclonal Anti-Caspase 9 antibody produced in mouse, ~2 mg/mL, clone CAS9, purified immunoglobulin, buffered aqueous solution
P248 Monoclonal Anti-PARP antibody produced in mouse, clone C-2-10, ascites fluid
WH0000142M1
Monoclonal Anti-PARP1 antibody produced in mouse, clone 3G4, purified immunoglobulin, buffered aqueous solution
SAB1403527
Monoclonal Anti-PARP1, (N-terminal) antibody produced in mouse, clone 2C7, purified immunoglobulin, buffered aqueous solution
PLA0184
Rabbit anti-PARP1 Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.