EMAIL THIS PAGE TO A FRIEND
Gene
Construction and characterization of a copy number-inducible fosmid library of Xanthomonas oryzae pathovar oryzae MAFF311018.
PMID 24835513
Abstract
A fosmid library of Xanthomonas oryzae pathovar oryzae MAFF311018 (T7174), the causative agent of bacterial blight on rice, was constructed and characterized. The average fosmid library insert size was >34kb, and 967 clones were uniquely positioned on its sequenced genome. The entire Xoo MAFF311018 genome was covered by end-sequenced clones with at least 5kb of overlap. The fosmid vector contains both the single-copy Escherichia coli fertility factor origin, which enhances fosmid stability, and the multi-copy IncPα origin, allowing amplification of copy number upon induction with l-arabinose. Real-time quantitative PCR on 12 randomly picked fosmid library clones determined that fosmid copy number increased 8- to 58-fold after 5hour induction. This library provides a new resource for complementation experiments and systematic functional studies in Xoo and related species.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A9524
DL-Arabinose, ≥98%
C5H10O5