EMAIL THIS PAGE TO A FRIEND
Journal of Chromatography A
Determination of bisphenol A, triclosan and their metabolites in human urine using isotope-dilution liquid chromatography-tandem mass spectrometry.
PMID 24835763
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
101052
2,3,4,5,6-Pentafluorobenzyl bromide, 99%
C7H2BrF5
516961
Ammonium formate, ≥99.995% trace metals basis
CH5NO2
09735
Ammonium formate, BioUltra, ≥99.0% (calc. based on dry substance, NT)
CH5NO2
70221
Ammonium formate, for mass spectrometry, ≥99.0%
CH5NO2
78314
Ammonium formate solution, BioUltra, 10 M in H2O
CH5NO2
714690
Ammonium formate solution, 10 mM in H2O, for HPLC
CH5NO2
09859
Ammonium hydroxide solution, BioUltra, ~1 M NH3 in H2O (T)
H5NO
40205
Ammonium hydroxide solution, ≥25% NH3 in H2O, semiconductor grade PURANAL (Honeywell 17814)
H5NO
40291
Ammonium hydroxide solution, ≥25% NH3 in H2O, semiconductor grade VLSI PURANAL (Honeywell 17605)
H5NO
40321
Ammonium hydroxide solution, ≥25% NH3 in H2O, semiconductor grade SLSI PURANAL (Honeywell 17301)
H5NO
01-4362
Ammonium hydroxide solution, 28% NH3 in H2O, suitable for determination of toxic metals
H5NO
D2625
Dansyl chloride, BioReagent, suitable for amino acid labeling, powder and chunks, ≥99% (HPLC)
C12H12ClNO2S
39220
Dansyl chloride, BioReagent, suitable for fluorescence, ≥99.0% (HPLC)
C12H12ClNO2S
03641
Dansyl chloride, for HPLC derivatization, ≥99.0% (HPLC)
C12H12ClNO2S
66742
Dichloromethane, Selectophore, ≥99.5%
CH2Cl2
02575
Dichloromethane, analytical standard
CH2Cl2
48538
Dichloromethane, analytical standard, stabilized
CH2Cl2
48600
Dichloromethane solution, certified reference material, 200 μg/mL in methanol
CH2Cl2
40042
Dichloromethane solution, certified reference material, 5000 μg/mL in methanol
CH2Cl2
52750
Hexane, analytical standard
C6H14
52765
Hexane, puriss. p.a., ACS reagent, ≥99.0% (GC)
C6H14
15671
Hexane, puriss., ≥95% (GC)
C6H14
M1550000
Methylene chloride, European Pharmacopoeia (EP) Reference Standard
CH2Cl2
PHR1557
Methylene Chloride, Pharmaceutical Secondary Standard; Certified Reference Material
CH2Cl2
33001
pentafluorobenzyl bromide (PFB-Br), analytical standard
C7H2BrF5
791776
Potassium carbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
CK2O3
P5833
Potassium carbonate, BioXtra, ≥99.0%
CK2O3
P1472
Potassium carbonate, meets USP testing specifications
CK2O3
367877
Potassium carbonate, 99.995% trace metals basis
CK2O3
590681
Potassium carbonate, anhydrous, powder, 99.99% trace metals basis
CK2O3