EMAIL THIS PAGE TO A FRIEND
The Science of the total environment
Occurrence of veterinary antibiotics and progesterone in broiler manure and agricultural soil in Malaysia.
PMID 24836135
Abstract
Repeated applications of animal manure as fertilizer are normal agricultural practices that may release veterinary antibiotics and hormones into the environment from treated animals. Broiler manure samples and their respective manure-amended agricultural soil samples were collected in selected locations in the states of Selangor, Negeri Sembilan and Melaka in Malaysia to identify and quantify veterinary antibiotic and hormone residues in the environment. The samples were analyzed using ultrasonic extraction followed by solid phase extraction (SPE) and liquid chromatography-tandem mass spectrometry (LC-MS/MS). The broiler manure samples were found to be contaminated with at least six target analytes, namely, doxycycline, enrofloxacin, flumequine, norfloxacin, trimethoprim and tylosin. These analytes were detected in broiler manure samples with maximum concentrations reaching up to 78,516 μg kg(-1) dry weight (DW) (doxycycline). For manure-amended agricultural soil samples, doxycycline and enrofloxacin residues were detected in every soil sample. The maximum concentration of antibiotic detected in soil was 1331 μg kg(-1) DW (flumequine). The occurrence of antibiotics and hormones in animal manure at high concentration poses a risk of contaminating agricultural soil via fertilization with animal manure. Some physico-chemical parameters such as pH, total organic carbon (TOC) and metal content played a considerable role in the fate of the target veterinary antibiotics and progesterone in the environment. It was suggested that these parameters can affect the adsorption of pharmaceuticals to solid environmental matrices.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A8523
Amoxicillin, potency: ≥900 μg per mg
C16H19N3O5S
1031503
Amoxicillin, United States Pharmacopeia (USP) Reference Standard
C16H19N3O5S · 3H2O
D9891
Doxycycline hyclate, ≥98% (HPLC)
C22H24N2O8 · HCl · 0.5H2O · 0.5C2H6O
33429
Doxycycline hyclate, VETRANAL, analytical standard
C22H24N2O8 · HCl · 0.5H2O · 0.5C2H6O
PHR1145
Doxycycline hyclate, Pharmaceutical Secondary Standard; Certified Reference Material
C22H24N2O8 · HCl · 0.5H2O · 0.5C2H6O
D3000000
Doxycycline hyclate, European Pharmacopoeia (EP) Reference Standard
C22H24N2O8 · HCl · 0.5H2O · 0.5C2H6O
1226003
Doxycycline hyclate, United States Pharmacopeia (USP) Reference Standard
C22H24N2O8 · HCl · 0.5H2O · 0.5C2H6O
17849
Enrofloxacin, ≥99.0% (HPLC)
C19H22FN3O3
33699
Enrofloxacin, VETRANAL, analytical standard
C19H22FN3O3
Y0001259
Enrofloxacin, European Pharmacopoeia (EP) Reference Standard
C19H22FN3O3
1235900
Enrofloxacin, United States Pharmacopeia (USP) Reference Standard
C19H22FN3O3
PHR1513
Enrofloxacin, Pharmaceutical Secondary Standard; Certified Reference Material
C19H22FN3O3
Y0001160
Enrofloxacin for system suitability, European Pharmacopoeia (EP) Reference Standard
C19H22FN3O3
E6376
Erythromycin, potency: ≥850 μg per mg
C37H67NO13
E5389
Erythromycin, BioReagent, suitable for cell culture
C37H67NO13
45674
Erythromycin, tested according to Ph.Eur.
C37H67NO13
PHR1039
Erythromycin, Pharmaceutical Secondary Standard; Certified Reference Material
C37H67NO13
E1300000
Erythromycin, for microbiological assay, European Pharmacopoeia (EP) Reference Standard
C37H67NO13
1242000
Erythromycin, United States Pharmacopeia (USP) Reference Standard
C37H67NO13
E0774
Erythromycin, meets USP testing specifications
C37H67NO13
45703
Erythromycin standard solution, 1 mg/mL in H2O
C37H67NO13
N9890
Norfloxacin, analytical standard, ≥98% (TLC)
C16H18FN3O3
33899
Norfloxacin, VETRANAL, analytical standard
C16H18FN3O3
N1230000
Norfloxacin, European Pharmacopoeia (EP) Reference Standard
C16H18FN3O3
Y0001301
Norfloxacin for peak identification, European Pharmacopoeia (EP) Reference Standard
C16H18FN3O3
Y0001015
Norfloxacin for system suitability, European Pharmacopoeia (EP) Reference Standard
C16H18FN3O3
T7883
Trimethoprim, ≥98% (HPLC)
C14H18N4O3
46984
Trimethoprim, VETRANAL, analytical standard
C14H18N4O3
92131
Trimethoprim, crystallized, ≥99.0% (HPLC)
C14H18N4O3
PHR1056
Trimethoprim, Pharmaceutical Secondary Standard; Certified Reference Material
C14H18N4O3